Assay Details
| Gene Symbol: pks1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157069.1 | NP_001150541.1 | 603 | 76 | ||
| GenBank mRNA | EU967359.1 | - | 603 | 76 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

pks1| Gene Symbol: pks1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157069.1 | NP_001150541.1 | 603 | 76 | ||
| GenBank mRNA | EU967359.1 | - | 603 | 76 | ||
Set Membership: |
|