Assay Details
| Gene Symbol: LOC100284209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157104.1 | NP_001150576.1 | 419 | 74 | ||
| GenBank mRNA | EU967508.1 | - | 419 | 74 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100284209| Gene Symbol: LOC100284209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157104.1 | NP_001150576.1 | 419 | 74 | ||
| GenBank mRNA | EU967508.1 | - | 419 | 74 | ||