Assay Details
| Gene Symbol: LOC100284726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157621.1 | NP_001151093.1 | 752 | 98 | ||
| GenBank mRNA | EU969472.1 | - | 752 | 98 | ||
More Information
Set Membership: |
|

LOC100284726
Genomic Map