Assay Details
| Gene Symbol: LOC100286267 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001159154.1 | NP_001152626.1 | 230 | 71 | ||
| GenBank mRNA | EU976514.1 | - | 230 | 71 | ||
More Information
Set Membership: |
|

LOC100286267
Genomic Map