Assay Details
| Gene Symbol: LOC100272398 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146878.1 | NP_001140350.1 | 139 | 65 | ||
| GenBank mRNA | BT038636.1 | - | 139 | 65 | ||
More Information
Set Membership: |
|

LOC100272398
Genomic Map