Assay Details
| Gene Symbol: IDP1632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001148111.1 | NP_001141583.1 | 1430 | 70 | ||
| GenBank mRNA | BT041656.1 | - | 1430 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IDP1632| Gene Symbol: IDP1632 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001148111.1 | NP_001141583.1 | 1430 | 70 | ||
| GenBank mRNA | BT041656.1 | - | 1430 | 70 | ||
Set Membership: |
|