Assay Details
Gene Symbol: LOC100274017 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001148402.1 | NP_001141874.1 | 1065 | 68 | ||
GenBank mRNA | BT042208.1 | - | 1065 | 68 | ||
BT084290.1 | - | 1092 | 68 | |||
EU967729.1 | - | 1051 | 68 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
