Assay Details
| Gene Symbol: LOC100274446 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001148805.1 | NP_001142277.1 | 517 | 79 | ||
| GenBank mRNA | BT035176.1 | - | 463 | 79 | ||
| BT043067.1 | - | 517 | 79 | |||
| BT060526.1 | - | 406 | 79 | |||
| EU976937.1 | - | 340 | 79 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the BT035176.1, BT060526.1 transcripts.
Set Membership: |
|

LOC100274446
Genomic Map