Assay Details
| Gene Symbol: LOC100279424 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001152434.1 | NP_001145906.1 | 556 | 76 | ||
| GenBank mRNA | BT054221.1 | - | 556 | 76 | ||
| EU970847.1 | - | 571 | 76 | |||
More Information
Set Membership: |
|

LOC100279424
Genomic Map