Assay Details
| Gene Symbol: LOC100280264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153192.1 | NP_001146664.1 | 1005 | 95 | ||
| GenBank mRNA | BT055882.1 | - | 1005 | 95 | ||
| HQ858734.1 | - | 823 | 95 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100280264| Gene Symbol: LOC100280264 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001153192.1 | NP_001146664.1 | 1005 | 95 | ||
| GenBank mRNA | BT055882.1 | - | 1005 | 95 | ||
| HQ858734.1 | - | 823 | 95 | |||