Assay Details
| Gene Symbol: LOC100281236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154155.1 | NP_001147627.1 | 629 | 82 | ||
| GenBank mRNA | EU956037.1 | - | 629 | 82 | ||
More Information
Set Membership: |
|

LOC100281236
Genomic Map