Assay Details
| Gene Symbol: LOC100281603 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154522.1 | NP_001147994.1 | 495 | 92 | ||
| GenBank mRNA | EU957222.1 | - | 495 | 92 | ||
More Information
Set Membership: | 
		   
					
						
							 | 
				

LOC100281603
Genomic Map