Assay Details
| Gene Symbol: LOC100282198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155110.1 | NP_001148582.1 | 1141 | 91 | ||
| GenBank mRNA | EU960004.1 | - | 1141 | 91 | ||
More Information
Set Membership: |
|

LOC100282198
Genomic Map