Assay Details
| Gene Symbol: LOC100282433 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155344.1 | NP_001148816.1 | 304 | 97 | ||
| GenBank mRNA | EU960879.1 | - | 304 | 97 | ||
More Information
Set Membership: |
|

LOC100282433
Genomic Map