Assay Details
| Gene Symbol: LOC100282599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155507.1 | NP_001148979.1 | 591 | 99 | ||
| GenBank mRNA | EU961586.1 | - | 591 | 99 | ||
More Information
Set Membership: |
|

LOC100282599
Genomic Map