Assay Details
Gene Symbol: LOC100283074 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001155976.1 | NP_001149448.1 | 296 | 56 | ||
GenBank mRNA | BT054999.1 | - | 248 | 56 | ||
EU963367.1 | - | 296 | 56 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
