Assay Details
| Gene Symbol: LOC100283478 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156378.1 | NP_001149850.1 | 536 | 93 | ||
| GenBank mRNA | BT065536.1 | - | 477 | 93 | ||
| EU964879.1 | - | 536 | 93 | |||
More Information
Set Membership: |
|

LOC100283478
Genomic Map