Assay Details
| Gene Symbol: LOC100283478 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156378.1 | NP_001149850.1 | 666 | 80 | ||
| GenBank mRNA | BT065536.1 | - | 607 | 80 | ||
| EU964879.1 | - | 666 | 80 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100283478| Gene Symbol: LOC100283478 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156378.1 | NP_001149850.1 | 666 | 80 | ||
| GenBank mRNA | BT065536.1 | - | 607 | 80 | ||
| EU964879.1 | - | 666 | 80 | |||