Assay Details
| Gene Symbol: LOC100283755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156654.1 | NP_001150126.1 | 318 | 62 | ||
| GenBank mRNA | EU965835.1 | - | 318 | 62 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100283755| Gene Symbol: LOC100283755 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156654.1 | NP_001150126.1 | 318 | 62 | ||
| GenBank mRNA | EU965835.1 | - | 318 | 62 | ||