Assay Details
| Gene Symbol: LOC100284924 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157819.1 | NP_001151291.1 | 602 | 63 | ||
| GenBank mRNA | EU970135.1 | - | 602 | 63 | ||
More Information
Set Membership: |
|

LOC100284924
Genomic Map