Assay Details
Gene Symbol: LOC100285159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001158053.1 | NP_001151525.1 | 349 | 96 | ||
GenBank mRNA | EU971058.1 | - | 349 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
