Assay Details
| Gene Symbol: LOC100286012 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158900.1 | NP_001152372.1 | 641 | 101 | ||
| GenBank mRNA | EU975181.1 | - | 641 | 101 | ||
More Information
Set Membership: |
|

LOC100286012
Genomic Map