Assay Details
| Gene Symbol: LOC100286085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158973.1 | NP_001152445.1 | 1333 | 74 | ||
| GenBank mRNA | EU975529.1 | - | 1333 | 74 | ||
More Information
Set Membership: |
|

LOC100286085
Genomic Map