Assay Details
Gene Symbol: LOC100286361 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001159248.1 | NP_001152720.1 | 157 | 110 | ||
GenBank mRNA | EU977008.1 | - | 157 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
