Assay Details
| Gene Symbol: LOC100272252 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146747.1 | NP_001140219.1 | 1195 | 79 | ||
| GenBank mRNA | BT038351.1 | - | 1195 | 79 | ||
More Information
Set Membership: |
|

LOC100272252
Genomic Map