Assay Details
| Gene Symbol: LOC100272293 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146780.1 | NP_001140252.1 | 1261 | 71 | ||
| GenBank mRNA | BT038428.1 | - | 1261 | 71 | ||
More Information
Set Membership: |
|

LOC100272293
Genomic Map