Assay Details
| Gene Symbol: LOC100273679 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001148091.1 | NP_001141563.1 | 671 | 70 | ||
| GenBank mRNA | BT041618.1 | - | 671 | 70 | ||
| EU967989.1 | - | 470 | 70 | |||
More Information
Set Membership: |
|

LOC100273679
Genomic Map