Assay Details
| Gene Symbol: chr101a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001148367.1 | NP_001141839.1 | 2360 | 81 | ||
| GenBank mRNA | BT042146.2 | - | 1107 | 81 | ||
| BT067025.1 | - | 2360 | 81 | |||
More Information
Set Membership: |
|

chr101a
Genomic Map