Assay Details
| Gene Symbol: LOC100281405 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154323.1 | NP_001147795.1 | 1662 | 90 | ||
| GenBank mRNA | BT061656.1 | - | 1662 | 90 | ||
| EU956607.1 | - | 1683 | 90 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100281405| Gene Symbol: LOC100281405 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154323.1 | NP_001147795.1 | 1662 | 90 | ||
| GenBank mRNA | BT061656.1 | - | 1662 | 90 | ||
| EU956607.1 | - | 1683 | 90 | |||