Assay Details
| Gene Symbol: LOC100282861 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155767.1 | NP_001149239.1 | 1182 | 91 | ||
| GenBank mRNA | EU962570.1 | - | 1182 | 91 | ||
More Information
Set Membership: |
|

LOC100282861
Genomic Map