Assay Details
| Gene Symbol: LOC100283039 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001155941.1 | NP_001149413.1 | 1535 | 95 | ||
| GenBank mRNA | BT067623.1 | - | 977 | 95 | ||
| BT069508.1 | - | 1535 | 95 | |||
| EU963245.1 | - | 989 | 95 | |||
More Information
Set Membership: |
|

LOC100283039
Genomic Map