Assay Details
| Gene Symbol: LOC100283150 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156052.1 | NP_001149524.1 | 655 | 85 | ||
| GenBank mRNA | BT037667.1 | - | 612 | 85 | ||
| EU963602.1 | - | 655 | 85 | |||
More Information
Set Membership: |
|

LOC100283150
Genomic Map