Assay Details
| Gene Symbol: LOC100283249 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156151.1 | NP_001149623.1 | 957 | 79 | ||
| GenBank mRNA | EU964003.1 | - | 957 | 79 | ||
More Information
Set Membership: |
|

LOC100283249
Genomic Map