Assay Details
| Gene Symbol: LOC100283352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156253.1 | NP_001149725.1 | 1434 | 75 | ||
| GenBank mRNA | EU964420.1 | - | 1434 | 75 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100283352| Gene Symbol: LOC100283352 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156253.1 | NP_001149725.1 | 1434 | 75 | ||
| GenBank mRNA | EU964420.1 | - | 1434 | 75 | ||