Assay Details
| Gene Symbol: LOC100283427 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156328.1 | NP_001149800.1 | 1248 | 89 | ||
| GenBank mRNA | EU964728.1 | - | 1248 | 89 | ||
More Information
Set Membership: |
|

LOC100283427
Genomic Map