Assay Details
| Gene Symbol: LOC100284174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157070.1 | NP_001150542.1 | 253 | 103 | ||
| GenBank mRNA | EU967362.1 | - | 253 | 103 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100284174| Gene Symbol: LOC100284174 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157070.1 | NP_001150542.1 | 253 | 103 | ||
| GenBank mRNA | EU967362.1 | - | 253 | 103 | ||