Assay Details
| Gene Symbol: cl405_1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157245.1 | NP_001150717.1 | 237 | 66 | ||
| GenBank mRNA | BT041654.1 | - | 216 | 66 | ||
| EU952816.1 | - | 133 | 66 | |||
| EU959833.1 | - | 213 | 66 | |||
| EU967995.1 | - | 237 | 66 | |||
More Information
Set Membership: |
|

cl405_1
Genomic Map