Assay Details
| Gene Symbol: cl405_1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157245.1 | NP_001150717.1 | 498 | 63 | ||
| GenBank mRNA | EU967995.1 | - | 498 | 63 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

cl405_1| Gene Symbol: cl405_1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157245.1 | NP_001150717.1 | 498 | 63 | ||
| GenBank mRNA | EU967995.1 | - | 498 | 63 | ||
Set Membership: |
|