Assay Details
| Gene Symbol: LOC100284461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157356.1 | NP_001150828.1 | 360 | 92 | ||
| GenBank mRNA | BT068120.1 | - | 366 | 92 | ||
| EU968458.1 | - | 360 | 92 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100284461| Gene Symbol: LOC100284461 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157356.1 | NP_001150828.1 | 360 | 92 | ||
| GenBank mRNA | BT068120.1 | - | 366 | 92 | ||
| EU968458.1 | - | 360 | 92 | |||