Assay Details
| Gene Symbol: LOC100284616 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157511.1 | NP_001150983.1 | 526 | 58 | ||
| GenBank mRNA | EU969050.1 | - | 526 | 58 | ||
More Information
Set Membership: |
|

LOC100284616
Genomic Map