Assay Details
| Gene Symbol: LOC100284836 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157731.1 | NP_001151203.1 | 1089 | 102 | ||
| GenBank mRNA | BT064920.1 | - | 1175 | 102 | ||
| EU969846.1 | - | 1089 | 102 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100284836| Gene Symbol: LOC100284836 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157731.1 | NP_001151203.1 | 1089 | 102 | ||
| GenBank mRNA | BT064920.1 | - | 1175 | 102 | ||
| EU969846.1 | - | 1089 | 102 | |||