Assay Details
| Gene Symbol: LOC100284914 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001157809.1 | NP_001151281.1 | 267 | 69 | ||
| GenBank mRNA | EU970105.1 | - | 267 | 69 | ||
| Gene Symbol: LOC100502306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001196784.1 | NP_001183713.1 | 465 | 69 | ||
| GenBank mRNA | BT087730.1 | - | 465 | 69 | ||
More Information
Set Membership: |
|

LOC100284914
Genomic Map