Assay Details
| Gene Symbol: LOC100285746 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001158637.1 | NP_001152109.1 | 436 | 87 | ||
| GenBank mRNA | EU973694.1 | - | 436 | 87 | ||
More Information
Set Membership: |
|

LOC100285746
Genomic Map