Assay Details
| Gene Symbol: LOC100273470 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001147907.1 | NP_001141379.1 | 545 | 79 | ||
| GenBank mRNA | BT038259.1 | - | 553 | 79 | ||
| BT041201.1 | - | 545 | 79 | |||
| EU962905.1 | - | 492 | 79 | |||
More Information
Set Membership: |
|

LOC100273470
Genomic Map