Assay Details
| Gene Symbol: LOC100281139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154058.1 | NP_001147530.1 | 718 | 57 | ||
| GenBank mRNA | EU955716.1 | - | 718 | 57 | ||
More Information
Set Membership: |
|

LOC100281139
Genomic Map