Assay Details
| Gene Symbol: LOC100281587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154506.1 | NP_001147978.1 | 2598 | 67 | ||
| GenBank mRNA | BT053799.1 | - | 2534 | 67 | ||
| EU957193.1 | - | 2598 | 67 | |||
More Information
Set Membership: |
|

LOC100281587
Genomic Map