Assay Details
| Gene Symbol: LOC100281677 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154597.1 | NP_001148069.1 | 1386 | 97 | ||
| GenBank mRNA | BT063192.1 | - | 1367 | 97 | ||
| EU957519.1 | - | 1386 | 97 | |||
More Information
Set Membership: |
|

LOC100281677
Genomic Map