Assay Details
| Gene Symbol: LOC100281677 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154597.1 | NP_001148069.1 | 1154 | 79 | ||
| GenBank mRNA | BT063192.1 | - | 1135 | 79 | ||
| EU957519.1 | - | 1154 | 79 | |||
More Information
Set Membership: |
|

LOC100281677
Genomic Map