Assay Details
| Gene Symbol: cl12627_1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001154996.1 | NP_001148468.1 | 1207 | 77 | ||
| GenBank mRNA | BT060587.1 | - | 1207 | 77 | ||
| EU959499.1 | - | 1210 | 77 | |||
More Information
Set Membership: |
|

cl12627_1
Genomic Map