Assay Details
| Gene Symbol: LOC100283223 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001156125.1 | NP_001149597.1 | 507 | 61 | ||
| GenBank mRNA | BT043085.1 | - | 471 | 61 | ||
| EU963883.1 | - | 507 | 61 | |||
More Information
Set Membership: |
|

LOC100283223
Genomic Map